Orders LinkContactUsLinkComponent
Bismuth Stain Kit, Electron Microscopy Sciences
Bismuth Stain Kit, Electron Microscopy Sciences
Bismuth Stain Kit 3
Catalog # 100503-420
Supplier #:  11436
Bismuth Stain Kit, Electron Microscopy Sciences
Catalog # 100503-420
CAS Number:  
Bismuth Stain Kit, Electron Microscopy Sciences
Bismuth Stain Kit 3
CAS Number:  
Price
$82.16
Log in for availability
Kit of 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Bismuth specifically stains Golgi beads and interchromatin granules after fixation with glutaraldehyde, whereas it stains nucleoli, basic protein, and biogenic amines after formaldehyde fixation
Bismuth staining of sections is relatively nonspecific: it can stain glycogen, lysosomes, ribosomes, polysaccharides, ferritin, and DNA. M.A. Hayat, "Basic Techniques for TEM" p. 184 (1986).
Specifications
Specifications
  • Description:
    Bismuth Stain Kit 3