Orders LinkContactUsLinkComponent
Clip-on Magnifiers, Electron Microscopy Sciences
Clip On Magnifier, 5, 203 mm (8")
  100502-160
 :  68260-05
Clip-on Magnifiers, Electron Microscopy Sciences
  100502-160
Clip-on Magnifiers, Electron Microscopy Sciences
Clip-on Magnifiers, Electron Microscopy Sciences
Clip On Magnifier, 5, 203 mm (8")
$83.83
Each
Clips on to any eyeglass frame leaving hands free.
Optical quality plastic lenses enables you to see tiny details without distortion using both eyes. Lightweight, comfortable and adjustable. Comes with carrying pouch.
  • Magnification:
    2.25×
  • Description:
    Clip On Magnifier
  • Diopters:
    5
  • Working distance:
    203 mm (8")