Orders LinkContactUsLinkComponent
Benchtop Mat, Electron Microscopy Sciences
Benchtop Mat, Electron Microscopy Sciences
Bench Top Mat Cut Size
Catalog # 100500-862
Supplier #:  64446
Benchtop Mat, Electron Microscopy Sciences
Catalog # 100500-862
Benchtop Mat, Electron Microscopy Sciences
Benchtop Mat, Electron Microscopy Sciences
Bench Top Mat Cut Size
Price
$579.76
Log in for availability
Box of 100
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A flexible polyethylene film backing which has a highly absorbent upper surface which traps water, dyes, resins, solvents, and radioactive solutions.
  • Sheets can be rolled up and discarded after use
  • Ideal for daily use in EM labs
Benchtop mat comes in a roll 50.8 cm (20") wide and 15.2 m (50') long in a dispenser carton, or a cut size of 20×24" sheets.
Specifications
  • Description:
    Bench Top Mat Cut Size