Orders LinkContactUsLinkComponent
NGW1 Graticles, Electron Microscopy Sciences
NGW1 Graticles, Electron Microscopy Sciences
NGW1 Graticule, 21 mm
Catalog # 100499-222
Supplier #:  68022-21
NGW1 Graticles, Electron Microscopy Sciences
Catalog # 100499-222
NGW1 Graticles, Electron Microscopy Sciences
NGW1 Graticles, Electron Microscopy Sciences
NGW1 Graticule, 21 mm
Price
$490.17
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Consists of 15 lines of equal length connecting the verticals of a regular hexagonal point network.
Reference: E.R.Weibel Lab. Invest. Vol. 22 pp 131-152. 'Principal and Methods for the Morphometric Study of the Lung and other Organs'.
Specifications
  • Description:
    NGW1 Graticule
  • Size:
    21 mm