Orders LinkContactUsLinkComponent
Petroff-Hausser Chambers, Electron Microscopy Sciences
Petroff Hausser chamber, 20 µm without grid
  100498-488
 :  63512-26
Petroff-Hausser Chambers, Electron Microscopy Sciences
  100498-488
Petroff-Hausser Chambers, Electron Microscopy Sciences
Petroff-Hausser Chambers, Electron Microscopy Sciences
Petroff Hausser chamber, 20 µm without grid
$1,863.62
Each
New improved Neubauer ruling pattern with 1/400 sq. mm of ruling. Designed for the counting of bacteria, sperm, blood platelets, and the content of vaccines.
Cell is 0.02 mm deep with 1.5 mm thick glass slide allows the use off dark field microscopy.
  • Description:
    Petroff Hausser chamber, 20 µm without grid