Inoculating Loops, Electron Microscopy Sciences
Product Details & Documents
Our high quality inoculating loops are made from platinum wire. The 15% iridium adds necessary rigidity permitting thinner wire for delicate work. The loop section is welded and smoothly finished to form a “perfect circle”.
Specifications
- Description:Inoculating Loops
- Size:3 x 45 mm
- Gauge:24 Gauge