Orders LinkContactUsLinkComponent
Standard Micro-Tool Sets, Electron Microscopy Sciences
Standard Micro-Tool Sets, Electron Microscopy Sciences
Microscopist's Set
Catalog # 100497-742
Supplier #:  62093-03
Standard Micro-Tool Sets, Electron Microscopy Sciences
Catalog # 100497-742
Standard Micro-Tool Sets, Electron Microscopy Sciences
Standard Micro-Tool Sets, Electron Microscopy Sciences
Microscopist's Set
Price
$801.75
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Micro-Needle Probe Set includes long Micro-needle, 90 degree Micro-needle, 45 degree Micro-needle, 90 degree Micro-hook, 180 degree Micro-hook, short Micro-needle, straight Ultra micro-needle, Micro-diamond-scriber, Micro-tool handle, and Micro-tool storage case.
Micro-Cutting Tool Set includes 3 sided Micro-graver, Micro-spade, oval Micro-graver, carbide Micro-scriber, Micro-tool storage case, Micro-chisel, Micro-knife 20 degree, Micro-scraper triangular, Micro-tool Handle.
Microscopist’s Set includes long Micro-needle, 20 degree Micro-knife, flexible Micro-spatula, 2-sided (metric & inch) Micro-scale, Micro-tool handle, Micro-chisel, Micro-diamond-scriber, 90 degree Micro-hook, straight Ultra-micro-needle, Micro-tool storage case.
Placement and Manipulating Set includes Micro-fork, 45 degree Micro-prober, Micro-spatula, 90 degree Micro-hook, Micro-tool handle, 10 mm Micro-scale, Micro-brush, Micro-manipulator, 30 degree bent Ultra-micro-needle, Micro-tool storage case.
Specifications
  • Description:
    Microscopist's Set