Orders LinkContactUsLinkComponent
Phillips® Screwdriver Set, Electron Microscopy Sciences
Phillips® Screwdriver Set, Electron Microscopy Sciences
Phillips Screwdriver, 2.0 mm, 4½"
Catalog # 100497-578
Supplier #:  62077-20
Phillips® Screwdriver Set, Electron Microscopy Sciences
Catalog # 100497-578
Phillips® Screwdriver Set, Electron Microscopy Sciences
Phillips® Screwdriver Set, Electron Microscopy Sciences
Phillips Screwdriver, 2.0 mm, 4½"
Price
$12.34
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A very handy set of Phillips screwdrivers which feature one of the smallest sizes available (1.20 mm). A knurled body with hexagonal heads and tempered steel blades.
Specifications
  • Description:
    Phillips Screwdriver
  • Length:
    4½"
  • Size:
    2.0 mm