Precision Pin Vises, Electron Microscopy Sciences
Product Details & Documents
These tool steel collets grip firmly when screw chucks are tightened.
- The chromium-plated handle is hollow which allows for the holding of long wires
The body is stepped, with the small portion knurled allowing for rotation between the fingers. The larger portion is smooth and it easily can be held firmly in lathes and drill presses.
Specifications
- Description:Precision pin vise, 37/8"