Orders LinkContactUsLinkComponent
Glennel Lapping Solutions, Electron Microscopy Sciences
Glennel Lapping Solutions, Electron Microscopy Sciences
Formula B Lube, 16 oz.
Catalog # 100497-398
Supplier #:  50376-10
Glennel Lapping Solutions, Electron Microscopy Sciences
Catalog # 100497-398
Glennel Lapping Solutions, Electron Microscopy Sciences
Glennel Lapping Solutions, Electron Microscopy Sciences
Formula B Lube, 16 oz.
Price
$61.08
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Penetrating formula designed to enhance cutting action and leaves a superior finish.
  • Formula C is made especially for oil soluble compounds
Glenoil Lube - Lapping vehicle is the ideal lap oil for all Glennel® UB Compounds and Diamond Sprays. It assures continued separation of diamond particles and constant removal of lap debris.
Formula B is a universal lapping vehicle, compatible with both water and oil soluble compounds. It is ideal in an environment where materials change constantly. This very versatile formulation allows you to change materials and compounds and still provides excellent lubricity and cutting action.
Specifications
  • Description:
    Formula B Lube
  • Size:
    16 oz.