Scintillation Vial Rack, Electron Microscopy Sciences
24, 11¾×5×3¾", Scintillation vial rack with handle
100496-346
: 72638
Welded polypropylene rack holds 20 ml scintillation vials.
- Autoclavable
- Resists nitric and hydrochloric acids
The hole measures ¹¹/₁₆" diameter.
- Description:Scintillation vial rack with handle
- No. of places:24
- W×D×H:11¾×5×3¾"