Orders LinkContactUsLinkComponent
Anionic Gold Tracer, Electron Microscopy Sciences
EM Anionic Gold Tracer, 5 ml, 6 nm
  100487-888
Anionic Gold Tracer, Electron Microscopy Sciences
  100487-888
 :  
Anionic Gold Tracer, Electron Microscopy Sciences
Anionic Gold Tracer, Electron Microscopy Sciences
EM Anionic Gold Tracer, 5 ml, 6 nm
 :  
$289.99
Each (5ml)
Anionic tracers are used to visualize positive charged moieties in specimens
  • Particle sizes of 0 8, 6, 10, 15 and 25 nm
  • Guaranteed shelf life of 18 months
  • Supplied in PBS with 15 mM NaN3
Aurion Gold Tracers are used to visualize charged moieties in specimens: anionic tracers bind to polycationic moieties (basic proteins e.g. histones), cationic tracers bind to polyanionic moieties (membranes, acidic proteins).
  • Description:
    EM Anionic Gold Tracer
  • Size:
    6 nm
  • Volume:
    5 ml