Anionic Gold Tracer, Electron Microscopy Sciences
Anionic tracers are used to visualize positive charged moieties in specimens
- Particle sizes of 0 8, 6, 10, 15 and 25 nm
- Guaranteed shelf life of 18 months
- Supplied in PBS with 15 mM NaN3
Aurion Gold Tracers are used to visualize charged moieties in specimens: anionic tracers bind to polycationic moieties (basic proteins e.g. histones), cationic tracers bind to polyanionic moieties (membranes, acidic proteins).
- Description:EM Anionic Gold Tracer
- Size:UltraSmall
- Volume:10 ml