Orders LinkContactUsLinkComponent
XRN-1
XRN-1
20 units
Catalog # 101449-366
Small Business Enterprise
Supplier #:  M0338S
XRN-1
Catalog # 101449-366
Small Business Enterprise
CAS Number:  
XRN-1
XRN-1
20 units
CAS Number:  
Price
$113.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
XRN-1 is highly processive 5´→3´ exoribonuclease, requiring 5´ monophosphate
It also acts on 5´ monophosphate ssDNA with greatly reduced efficiency. XRN-1 is not efficient in removing RNAs with recessed 5´P, like tRNA. It does not cleave dsDNA, ssDNA and RNA which contain di, triphosphate, OH and capped 5´ ends.
Specifications
Specifications
  • Size:
    20 units
  • Cat. No.:
    101449-366
  • Enzyme Name:
    XRN-1