Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
CHES/ Sodium hydroxide, buffer, Rigaku

CHES/ Sodium hydroxide, buffer, Rigaku

CHES/NaOH, 0.5M, 100 mL, 10.8
Catalog # 101443-264
Marketsource
Supplier:   Rigaku Reagents
Supplier #:  1008022
CHES/ Sodium hydroxide, buffer, Rigaku
Catalog # 101443-264
Supplier:  Rigaku Reagents
Marketsource
CAS Number:  
CHES/ Sodium hydroxide, buffer, Rigaku
CHES/ Sodium hydroxide, buffer, Rigaku

CHES/NaOH, 0.5M, 100 mL, 10.8

CAS Number:  
Price
$94.15
Log in for availability
Each (100ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Caution: Intended for Research and Development Use Only.

Specifications

Specifications

  • Description:
    CHES/NaOH, 0.5M
  • Size:
    100 mL
  • pH:
    10.8

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.