Needle and Knife Interchangeable Handle, Electron Microscopy Sciences
Handle L x Diameter: 92 x 6 mm (3 5/8 x 1/4")
Made from stainless steel, pin-vise type Handle, which accepts either dissecting needles or a knife.
Handle L x Diameter: 92 x 6 mm (3 5/8 x 1/4")
Needle: available with straight or bent, 60 mm (2 1/2") long
Knife: spear type with single or double edge, 75 mm (3") long
- Description:Spear Shape Knife, Single Edge, 3