Orders LinkContactUsLinkComponent
Manomark Pen All Surface Marker, Electron Microscopy Sciences
Manomark Pen All Surface Marker, Electron Microscopy Sciences
Manomark pen, Black
Catalog # 101413-506
Supplier #:  72127-B1
Manomark Pen All Surface Marker, Electron Microscopy Sciences
Catalog # 101413-506
Manomark Pen All Surface Marker, Electron Microscopy Sciences
Manomark Pen All Surface Marker, Electron Microscopy Sciences
Manomark pen, Black
Price
$18.25
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Writes on all plastic, glass, metal, wool, paper and cloth, including "PTFE", fluoropolymer resins
  • With its precision seal valve the pens life is preserved and as well it allows for a smooth flow of ink even if it’s left uncapped
  • Once dry, the solvent base ink is temperature resistant from 0 to 155 °C (32 to 310 °F)
Markings are insoluble in water or hydrocarbons, resist many solvents, and are easily removed with alcohol.
Specifications
  • Color:
    Black
  • Description:
    Manomark pen