Orders LinkContactUsLinkComponent
Fluid Dispenser Bottles, Electron Microscopy Sciences
Fluid Dispenser Bottles, Electron Microscopy Sciences
Fluid Dispenser, 180 ml Ea
Catalog # 101412-924
Supplier #:  64299-02
Fluid Dispenser Bottles, Electron Microscopy Sciences
Catalog # 101412-924
Fluid Dispenser Bottles, Electron Microscopy Sciences
Fluid Dispenser Bottles, Electron Microscopy Sciences
Fluid Dispenser, 180 ml Ea
Price
$53.86
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These ESD safe polyethylene solvent dispensers are available in three standard sizes.
  • Surface resistivity of 10E7 to 10E9
These dispensers have neither a stopper nor a cap to remove. Just touch the dispensing dish with a brush cloth or applicator and solvent is brought to the surface instantly as needed. Stainless steel valves seal contents in bottle until used. The fluid pumps into the dish and it does not drip back into the bottle, keeping stored liquids uncontaminated.
Specifications
  • Description:
    Fluid Dispenser, 180 ml Ea