Orders LinkContactUsLinkComponent
Bacti-Cinerator IV, Electron Microscopy Sciences
Replacement Bacti-Cinerator III Heat Element, 240 V
  101412-634
 :  62420-54
Bacti-Cinerator IV, Electron Microscopy Sciences
  101412-634
Bacti-Cinerator IV, Electron Microscopy Sciences
Bacti-Cinerator IV, Electron Microscopy Sciences
Replacement Bacti-Cinerator III Heat Element, 240 V
$558.10
Each
The Baci-Cinerator IV sterilizes by infrared heat (Heater Element contains no asbestos) - organic material is incinerated deep inside the ceramic tunnel. To use, simply insert loop or needle into the tunnel. Complete sterilization takes only 5 to 7 seconds at optimum sterilizing temperature of 1500 °F.
  • Prevents infectious spatter and/or cross contamination
  • No open flame -no hazardous gas
  • No need to wait for the loops or needles to turn red
  • Does not require oxygen so it can be used in anaerobic chambers
  • UL approved
  • Description:
    Replacement Bacti-Cinerator III Heat Element
  • Electrical:
    240 V
Accessories
  •  
    Description
  • 100498-070
    Replacement Switch, Bacti-Cinerator III
    Each - $114.02