Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
FORM - NON-HAZARDOUS WASTE FOR
Catalog # 101378-262
Supplier:
Label Master
Expand image
FORM - NON-HAZARDOUS WASTE FOR
Catalog # 101378-262
Supplier:
Label Master
Supplier Number:
CF14