Orders LinkContactUsLinkComponent
Atomizer Rubber Bulb, Electron Microscopy Sciences
Atomizer Rubber Bulb, Electron Microscopy Sciences
Atomizer Rubber Bulb, 75cc
Catalog # 100505-048
Supplier #:  60804
Atomizer Rubber Bulb, Electron Microscopy Sciences
Catalog # 100505-048
Atomizer Rubber Bulb, Electron Microscopy Sciences
Atomizer Rubber Bulb, Electron Microscopy Sciences
Atomizer Rubber Bulb, 75cc
Price
$27.35
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A natural rubber bulb with 4" tubing on one end. Used for dusting surfaces which should not be wiped.
A two-way valve is inserted into one end to prevent suck back and a rubber nozzle prevents scratches on the surface that have to be cleaned. This device also reduces the need for freon dusters.
Specifications
  • Description:
    Atomizer Rubber Bulb, 75cc