Glass Microbeakers, Electron Microscopy Sciences
Product Details & Documents
Used for the processing of micro-samples and for visual or microscopic examination, staining, fixation, infiltration, liquid evaporation, bacterial growth, weighing, etc.
Specifications
- Ext.Ø:25 mm
- Capacity:5 ml
- Height:19 mm