Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

PTFE Beakers, Electron Microscopy Sciences

50 ml, PTFE Beakers, White, 58 mm, 38 mm
  100503-244
PTFE Beakers, Electron Microscopy Sciences
  100503-244
PTFE Beakers, Electron Microscopy Sciences
PTFE Beakers, Electron Microscopy Sciences

50 ml, PTFE Beakers, White, 58 mm, 38 mm

 
$54.92
   
Each
 
   
 

These beakers are statically molded from virgin "PTFE" and machine finished to give a smooth surface for ease of cleaning and to eliminate contamination.

  • With pouring spout
  • Virtually immune to chemical attack

Working temperature range is from cryogenic to 260°C (500°F). Ideal for use in wafer etching.

 
  • Ext.Ø:
    38 mm
  • Capacity:
    50 ml
  • Color:
    White
  • Description:
    PTFE Beakers
  • Height:
    58 mm

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.