Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Bouin's Solution, Electron Microscopy Sciences

Bouin's Solution, Electron Microscopy Sciences

Bouin's Solution, 1 L
Catalog # 100503-022
Minority or Woman-Owned Business Enterprise
Supplier #:  15990-01
Bouin's Solution, Electron Microscopy Sciences
Catalog # 100503-022
Minority or Woman-Owned Business Enterprise
CAS Number:  
Bouin's Solution, Electron Microscopy Sciences
Bouin's Solution, Electron Microscopy Sciences

Bouin's Solution, 1 L

CAS Number:  
Price
$105.29
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Bouin's fixative is used on biopsy specimens of the gastrointestinal tract.

  • Used on biopsy specimens
  • Fixation and staining fluid

Bouin's Solution is used as a fixation and staining fluid. In prefilled leak-proof histo-containers it is ready-to-use. Formulated with Picric Acid saturated aqueous solution 75%, Formalin 25% and Glacial Acetic acid 5% in de-ionized water.
Bouin's fixative is excellent for use on biopsy specimens of the gastrointestinal tract. Tissue from the endocrine system are well fixed and many antibodies react well with tissue fixed in Bouin's. Bouin's solution may be used as a routine fixative or a staining solution, but should not be used as a preservative for tissue. Tissue cannot be held in Bouin's solution for extended periods. The maximum fixation time in this solution should be less than 48 hours and should be transferred to alcohol.

Specifications

Specifications

  • Description:
    Bouin's Solution
  • Size:
    1 L
  • Boiling Point:
    -21 °C (-6 °F)
  • Cat. No.:
    100503-022
  • Flash Point:
    0 °C (32 °F)
  • Density:
    0.99507 g/cm³ (8.304 lbs/gal)
  • Storage Temperature:
    RT

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.