Orders LinkContactUsLinkComponent
All Plastic Syringes, Electron Microscopy Sciences
All Plastic Syringes, Electron Microscopy Sciences
Syringe 30 cc, Clear, Eccentric
Catalog # 100500-896
All Plastic Syringes, Electron Microscopy Sciences
Catalog # 100500-896
Supplier:  
All Plastic Syringes, Electron Microscopy Sciences
All Plastic Syringes, Electron Microscopy Sciences
Syringe 30 cc, Clear, Eccentric
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
SYRINGE ST SLIP TIP 30ML PK56 Catalog #BD302833
Product BD302833 is similar to 100500-896 but manufacturer and specifications may vary.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All plastic syringes are the choice for any situation needing an inert, hypoallergenic syringe. Because of their purity, they are indicated for IVF, embryo transfer, chromatography and many laboratory procedures.
  • Latex free and contain no rubber, no silicone oil or styrene
  • Manufactured only from laboratory grade polypropylene and polyethylene
  • Unique plastic syringes have a positive safety stop to prevent accidental spills
Packaging: These syringes are individually sterile strip packed.
Specifications
  • Type:
    Eccentric
  • Color:
    Clear
  • Description:
    Syringe 30 cc