Orders LinkContactUsLinkComponent
Kendall Treated Pipette Tips, Electron Microscopy Sciences
Kendall Treated Pipette Tips, Electron Microscopy Sciences
Universal Tips, Not Treated, 5 - 200 µl, Yellow, 10 Racks/Case, 96 Tips/Quick Rack
Catalog # 100500-162
Supplier #:  72446-05
Kendall Treated Pipette Tips, Electron Microscopy Sciences
Catalog # 100500-162
Kendall Treated Pipette Tips, Electron Microscopy Sciences
Kendall Treated Pipette Tips, Electron Microscopy Sciences
Universal Tips, Not Treated, 5 - 200 µl, Yellow, 10 Racks/Case, 96 Tips/Quick Rack
Price
$80.58
Log in for availability
Case of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
KENDALL’s low binding tips ensure accuracy and precision by bonding a proprietary polymer to the resin. Precision and accuracy are increasingly important in molecular biology labs. A pipetter can be 99% accurate and precise, but the tip can com-promise performance by retaining or binding a sample.
  • Not detectable/Non-reactive
  • Extracted only by a fluorinated solvent
  • Autoclaving will not compromise performance
  • Tips and racks are fully autoclavable
  • Graduation marks at 2 and 5 µl
  • Yellow tips non-graduated, not-treated
  • Available bulk, racked, racked sterile, EcoStack racks and QuickRack tip reloading system
Reduces the surface energy of liquid and repels any substance. Extended length reaches bottom of tubes without contaminating the pipettor shaft. Low Binding Polymer Technology: A manufacturing process that binds a perfluorooctyl polymer to polypropylene resin–resulting in an exposed trifluoromethyl surface.
Graduations:
0.1 to 10 µl: 2 and 5 µl
5 to 200 µl / 5 to 300 µl / 101 to 1000 µl: 10, 50 and 100 µl
Available in convenient hinged racked package, with and without low binding surface. Available bulk (not treated), racked and racked sterile (low binding).
Available in QuickRack
0.5 to 10 µl Crystal/Eppendorf Tips-Natural: Fits Eppendorf ultra micro pipetters
1 to 200 µl Universal Tips: Universal fit for research-grade pipetters
P2/P10 Ultra Micro Tip – Natural: Universal fit for ultra-micro pipetters
Specifications
  • Volume:
    5 - 200 µl
  • Tip color:
    Yellow
  • Packed:
    10 Racks/Case, 96 Tips/Quick Rack
  • Tip style:
    Universal Tips, Not Treated