Orders LinkContactUsLinkComponent
Chessboard Squares Graticules, Electron Microscopy Sciences
Chessboard Squares Graticules, Electron Microscopy Sciences
NE15 Eyepiece Scale, 19 mm
Catalog # 100499-124
Supplier #:  68008-19
Chessboard Squares Graticules, Electron Microscopy Sciences
Catalog # 100499-124
Chessboard Squares Graticules, Electron Microscopy Sciences
Chessboard Squares Graticules, Electron Microscopy Sciences
NE15 Eyepiece Scale, 19 mm
Price
$466.08
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The dark squares are used as an alternative to simple grids for area of specimen determination and paricle counting.
  • Translucent
  • Alternative light and dark squares help to reduce eyestrain
Specifications
  • Description:
    NE15 Eyepiece Scale
  • Size:
    19 mm