Chessboard Squares Graticules, Electron Microscopy Sciences
Product Details & Documents
The dark squares are used as an alternative to simple grids for area of specimen determination and paricle counting.
- Translucent
- Alternative light and dark squares help to reduce eyestrain
Specifications
- Description:NE15 Eyepiece Scale
- Size:16 mm