Orders LinkContactUsLinkComponent
Sterilizing Tray Plastic, Electron Microscopy Sciences
Sterilizing Tray Plastic, Electron Microscopy Sciences
Sterilization Tray with Silicone Mat and Lid, 191 x 102 x 38 mm
Catalog # 100498-920
Supplier #:  66114-02
Sterilizing Tray Plastic, Electron Microscopy Sciences
Catalog # 100498-920
Sterilizing Tray Plastic, Electron Microscopy Sciences
Sterilizing Tray Plastic, Electron Microscopy Sciences
Sterilization Tray with Silicone Mat and Lid, 191 x 102 x 38 mm
Price
$233.12
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Perforated sterilizing tray has a silicone mat which provides flexibility int the layout the instruments for sterilization. A unique "grid" system exists in the base, tray and lid, making it easy to install the mat required, protecting your instruments properly.
  • Made of high strength Ultem® Plastic
  • Autoclave, Steam, Dry Heat, and Chemical Sterilization are possible
  • Has an extended Life Cycle
  • Trays are stackable
  • Economical and Flexible Design
  • Bases have pebble-type surface to keep mats from sticking
The trays are stackable and they have integral feet in base and corner ridges on lids which allow for easier handling in the autoclave and for storage.
Specifications
  • Description:
    Sterilization Tray with Silicone Mat and Lid
  • Size:
    191 x 102 x 38 mm