Orders LinkContactUsLinkComponent
Inoculating Loops, Electron Microscopy Sciences
Inoculating Loops, 5 x 45 mm, 24 Gauge
  100498-086
 :  62433-05
Inoculating Loops, Electron Microscopy Sciences
  100498-086
Inoculating Loops, Electron Microscopy Sciences
Inoculating Loops, Electron Microscopy Sciences
Inoculating Loops, 5 x 45 mm, 24 Gauge
$344.61
Each
Our high quality inoculating loops are made from platinum wire. The 15% iridium adds necessary rigidity permitting thinner wire for delicate work. The loop section is welded and smoothly finished to form a “perfect circle”.
  • Description:
    Inoculating Loops
  • Size:
    5 x 45 mm
  • Gauge:
    24 Gauge