Orders LinkContactUsLinkComponent
Diamond Points, Electron Microscopy Sciences
Diamond Points, Electron Microscopy Sciences
Diamond Points, Tapered, 5.2 mm, Cone
Catalog # 100497-984
Supplier #:  62155-52
Diamond Points, Electron Microscopy Sciences
Catalog # 100497-984
Diamond Points, Electron Microscopy Sciences
Diamond Points, Electron Microscopy Sciences
Diamond Points, Tapered, 5.2 mm, Cone
Price
$41.63
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All diamond points are mounted on 2.35 mm (3/32”) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
Specifications
  • Ø:
    5.2 mm
  • Description:
    Diamond Points, Tapered
  • Shape:
    Cone