Orders LinkContactUsLinkComponent
Diamond Points, Electron Microscopy Sciences
Diamond Points, Narrow, 1.8 mm, Cone
  100497-972
 :  62155-18
Diamond Points, Electron Microscopy Sciences
  100497-972
Diamond Points, Electron Microscopy Sciences
Diamond Points, Electron Microscopy Sciences
Diamond Points, Narrow, 1.8 mm, Cone
$37.91
Each
All diamond points are mounted on 2.35 mm (3/32”) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
  • Ø:
    1.8 mm
  • Description:
    Diamond Points, Narrow
  • Shape:
    Cone