Vacuum Mounting Blocks, Electron Microscopy Sciences
Product Details & Documents
Vacuum Mounting Bocks - Vacuum Plates for use with #50151-00
Specifications
- Description:2.125" (54mm) Diameter Mounting Block; 2" (50mm) Diameter Vacuum Pattern.