Orders LinkContactUsLinkComponent
MAC X-ray Microanalysis NBS Standards, Electron Microscopy Sciences
MAC X-ray Microanalysis NBS Standards, Electron Microscopy Sciences
NBS SRM 481
Catalog # 100495-238
Supplier #:  80076-01
MAC X-ray Microanalysis NBS Standards, Electron Microscopy Sciences
Catalog # 100495-238
CAS Number:  
MAC X-ray Microanalysis NBS Standards, Electron Microscopy Sciences
NBS SRM 481
CAS Number:  
Price
$2,504.02
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All MAC X-ray Microanalysis standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems. All of our standards are polished to a 0.25 micron diamond finish and carbon coated. They are supplied with fully authenticated certificates of analyses and a location map for standard identification.
NBS glass SRM 481
NBS glass
SRM 482 Au/Cu Set of 6 Standards, comprises 4 alloys and 2 pure metals.
NBS Standard SRM 1872
Set of 3 glasses K-453, K491, K968.
Specifications
Specifications
  • Description:
    NBS SRM 481