Orders LinkContactUsLinkComponent
Five-Channel Timer, Electron Microscopy Sciences
Five-Channel Timer, Electron Microscopy Sciences
Timer 5-channel certified
Catalog # 100493-814
Supplier #:  72600-02
Five-Channel Timer, Electron Microscopy Sciences
Catalog # 100493-814
Five-Channel Timer, Electron Microscopy Sciences
Five-Channel Timer, Electron Microscopy Sciences
Timer 5-channel certified
Price
$130.67
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Timer has four independent channels, each of which counts up or down for any interval from 1 second to 100 hours.
  • Memory recalls last timer setting for reuse
  • Has a clock mode
Its extra large display indicates channels that are in use. Each one has a distinctive 1 minute alarm. Automatically begins to count up to indicate time elapsed after count down reaches zero.
Dimensions: 2⁷/₈×2¹/₂×⁹/₁₆"
Weight: 2 oz.
Delivery information: Comes complete with a stand, pocket clip, and magnet.
Specifications
  • Description:
    Timer 5-channel certified