Orders LinkContactUsLinkComponent
Film Casting Device, Electron Microscopy Sciences
Film Casting Device, Electron Microscopy Sciences
Film Casting Device Set
Catalog # 100493-270
Supplier #:  71305-01
Film Casting Device, Electron Microscopy Sciences
Catalog # 100493-270
Film Casting Device, Electron Microscopy Sciences
Film Casting Device, Electron Microscopy Sciences
Film Casting Device Set
Price
$1,462.73
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
An all glass apparatus that casts uniformly thin films of parlodion, formvar, or butvar directly onto 1 x 3 microscope slides.
  • Film casting solution can be used repeatedly
  • Unit requires 100 mL of film casting solution to start
A built-in fine-pressure-release valve helps control the speed of drainage. The thickness of the film is controlled by the concentration of the film solution and the rate of the drainage.
Specifications
  • Description:
    Film Casting Device Set