Orders LinkContactUsLinkComponent
Dissecting Needles, Electron Microscopy Sciences
Dissecting Needles, Electron Microscopy Sciences
Single Edge Dissecting Needle
Catalog # 100492-042
Supplier #:  72946
Dissecting Needles, Electron Microscopy Sciences
Catalog # 100492-042
Dissecting Needles, Electron Microscopy Sciences
Dissecting Needles, Electron Microscopy Sciences
Single Edge Dissecting Needle
Price
$12.39
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The stainless steel with spear shaped head measures 127 mm (5") long. The wooden handle has a needle tip.
Specifications
  • Description:
    Single Edge Dissecting Needle