Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Dust-Off Plus Model (DPN), Electron Microscopy Sciences

Dust-Off Plus Model (DPN), Electron Microscopy Sciences

Dust-Off Plus Refill, DPNR, 227 g (8 oz.)
Catalog # 100491-660
Supplier #:  70847
Dust-Off Plus Model (DPN), Electron Microscopy Sciences
Catalog # 100491-660
Dust-Off Plus Model (DPN), Electron Microscopy Sciences
Dust-Off Plus Model (DPN), Electron Microscopy Sciences

Dust-Off Plus Refill, DPNR, 227 g (8 oz.)

Price
$45.42
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Contains formula N (tetrafluoroethane).

  • 100% safe for the ozone
  • Contains no flammable elements

It is provided with a patented 360° vector valve. The multi-directional valve is used for easy blasting in hard to reach areas. The valve can be removed from the can anytime without any loss of gas.

Delivery information: The kit consists of a 8 oz. can and a vector valve nozzle.

Specifications
  • Volume:
    227 g (8 oz.)
  • Description:
    Dust-Off Plus Refill, DPNR

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.