Orders LinkContactUsLinkComponent
Tray Set with Plastic Boxes, Electron Microscopy Sciences
Tray Box
  100490-578
 :  64348-05
Tray Set with Plastic Boxes, Electron Microscopy Sciences
  100490-578
Tray Set with Plastic Boxes, Electron Microscopy Sciences
Tray Set with Plastic Boxes, Electron Microscopy Sciences
Tray Box
$145.96
Pack of 5
A compact, solid fiberboard tray box makes instituting a plastic box/sample storage system more convenient and efficient.
  • Available with or without accompanying plastic boxes
To form a tray set, use plastic boxes for specimen storage, and then organize boxes within the fiberboard tray Box. These sets eliminates misplacement or inadvertent sample loss and allows for rapid retrieval of specimens.
  • Description:
    Tray Box