Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
LEO SEM Mount, Electron Microscopy Sciences

LEO SEM Mount, Electron Microscopy Sciences

LEO Microscope Mounts Slotted Head, 500
Catalog # 100488-664
Supplier #:  75192
LEO SEM Mount, Electron Microscopy Sciences
Catalog # 100488-664
LEO SEM Mount, Electron Microscopy Sciences
LEO SEM Mount, Electron Microscopy Sciences

LEO Microscope Mounts Slotted Head, 500

Price
$480.81
Log in for availability
Pack of 500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

These mounts with tapered end pin

  • Slotted head 12.7 mm (1/2") diameter
  • Pin 3.1 x 5 mm (1/8 x 1/4")
Specifications
  • Description:
    LEO Microscope Mounts Slotted Head
  • Size:
    500

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.