SRT® SEC HPLC Column, Sepax Technologies
Product Details & Documents
SRT® SEC phase, developed based on innovative surface coating technology, comprises of uniform, hydrophilic, and neutral nanometer thick film chemically bonded to high purity and mechanically stabilized silica.
- High separation efficiency and resolution
- High capacity
- High lot-to-lot reproducibility
- High protein recovery with intact biological activity
- Negligible non-specific interactions
SRT® uses stand-up monolayer on porous silica as oppose to SRT®-C of lay-down monolayer, offers ideal phase for monoclonal antibodies, proteins, peptides, nucleic acids, oligonucleotides, virus and water-soluble polymers.
Specifications
- Column Format:Analytical
- Ø int.:4.60 mm
- Particle Size:5 µm
- Pore size:300 Å
- Length:300 mm
- Packing material:Chemically bonded and hydrophilic silica
- Phase:SRT® SEC
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



