Graduated Transfer Pipettes, Electron Microscopy Sciences
Product Details & Documents
These disposable graduated transfer pipettes are made up of polyethylene.
- Low-affinity surface
- Attached bulb avoids cross-contamination
- One step biohazard disposable
These pipettes are ideal for clinical applications, blood banking, remove, disperse small volume, transfer in and out of multi well plates, reagent handing, pH meter maintenance, etc.
Specifications
- Ø×L:7.9×15.5 cm
- Capacity:7.5 ml
- Description:Graduated Transfer Pipettes
- Packed:Individually Packed
- Graduations:3 mL
- Bulb draw:3.2 ml
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



