Toren's Method for Mast Cells, Electron Microscopy Sciences
Pollack's Trichrome Solution in 50% Alcohol, 3A, 100 ml
Catalog # 100504-650
Supplier: Electron Microscopy Sciences
Supplier #: 26472-03
Toren's Method for Mast Cells, Electron Microscopy Sciences
Catalog # 100504-650
Supplier: Electron Microscopy Sciences
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Description:Pollack's Trichrome Solution in 50% Alcohol, 3A
- Size:100 ml
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



