Vial Rack, Electron Microscopy Sciences
Product Details & Documents
This rugged polypropylene rack holds 48 vials, size 1 dram (15 x 45 mm); or 4 ml tissue culture vials; or 2 mL serum vials in 4 x 12 configuration with alpha-numeric indexing.
The rack is constructed from chemically resistant, autoclavable (121 °C for 20 minutes) polypropylene and features corner posts to assure stability when stacking empty.
Specifications
- Description:Rack for 1 Dram Shell Vial
- Dimensions:94 x 266 x 28 mm (3 3/4 x 10 5/8 x 1 1/8")