Orders LinkContactUsLinkComponent
Paterson® Super System 4 Developing Tanks, Electron Microscopy Sciences
Paterson® Super System 4 Developing Tanks, Electron Microscopy Sciences
Paterson Universal Tank & Reel
Catalog # 100501-472
Supplier #:  74342-15
Paterson® Super System 4 Developing Tanks, Electron Microscopy Sciences
Catalog # 100501-472
Paterson® Super System 4 Developing Tanks, Electron Microscopy Sciences
Paterson® Super System 4 Developing Tanks, Electron Microscopy Sciences
Paterson Universal Tank & Reel
Price
$64.89
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Super System 4 is simpler to load, faster to fill, and easier to clean than any other tank. It has a large diameter one-piece lid and funnel, which clips into place positively and securely enabling the tank to be filled and emptied quickly. The Paterson 35 mm tank takes a single 35 mm film. The Paterson Universal tank takes two 35 mm films or a single 120/220 film. Supplied complete with one 35mm/120/220 Auto Load Reel, Watertight EVA cap prevents leakage during inversion agitation. Uses less than 300 mL solution for a single 35 mm film.
Specifications
  • Description:
    Paterson Universal Tank & Reel