Orders LinkContactUsLinkComponent
Coverglass Staining Jars w/Plastic Caps, Electron Microscopy Sciences
Coverglass Staining Jars w/Plastic Caps, Electron Microscopy Sciences
Coverglass Staining Jars with Screw Caps
Catalog # 100501-230
Supplier #:  72242-24
Coverglass Staining Jars w/Plastic Caps, Electron Microscopy Sciences
Catalog # 100501-230
Coverglass Staining Jars w/Plastic Caps, Electron Microscopy Sciences
Coverglass Staining Jars w/Plastic Caps, Electron Microscopy Sciences
Coverglass Staining Jars with Screw Caps
Price
$295.55
Log in for availability
Pack of 4
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
For use with cover glass sizes 18 to 22 mm, accommodating up to eight back-to-back.
Plastic screw caps have been tested to be resistant against most staining solutions.
Specifications
  • Description:
    Coverglass Staining Jars with Screw Caps