Orders LinkContactUsLinkComponent
Square and Grid Scales, Electron Microscopy Sciences
NE11 Eyepiece Scales, 21 mm, Grid (net) 1.00 mm pitch
  100499-174
 :  68015-21
Square and Grid Scales, Electron Microscopy Sciences
  100499-174
Square and Grid Scales, Electron Microscopy Sciences
Square and Grid Scales, Electron Microscopy Sciences
NE11 Eyepiece Scales, 21 mm, Grid (net) 1.00 mm pitch
$240.22
Each
Simple grids are convenient for making sketches of the obseved speimen on graph paper.
They are also useful for particle counting.
  • Description:
    NE11 Eyepiece Scales
  • Size:
    21 mm
  • Divisions:
    Grid (net) 1.00 mm pitch