Orders LinkContactUsLinkComponent
Indexed Grid Scales, Electron Microscopy Sciences
Indexed Grid Scales, Electron Microscopy Sciences
NE35 Graticule, 19 mm
Catalog # 100499-166
Supplier #:  68014-19
Indexed Grid Scales, Electron Microscopy Sciences
Catalog # 100499-166
Indexed Grid Scales, Electron Microscopy Sciences
Indexed Grid Scales, Electron Microscopy Sciences
NE35 Graticule, 19 mm
Price
$233.47
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Useful for area of specimen determinations, especially rectangular shapes, also for particle counting.
Specifications
  • Description:
    NE35 Graticule
  • Size:
    19 mm