Orders LinkContactUsLinkComponent
Bright-Line Counting Chamber, Electron Microscopy Sciences
Bright-Line Counting Chamber, Electron Microscopy Sciences
Hemacytometer Cover Glass, 0.4 mm (1461)
Catalog # 100498-504
Supplier #:  63514-30
Bright-Line Counting Chamber, Electron Microscopy Sciences
Catalog # 100498-504
Bright-Line Counting Chamber, Electron Microscopy Sciences
Bright-Line Counting Chamber, Electron Microscopy Sciences
Hemacytometer Cover Glass, 0.4 mm (1461)
Price
$72.56
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
For accurate counting of blood cells platelets, dust/yeast cluster and specimen samples. Sharp contrast from rhodium plated on "V-Load". Triple dividing lines clearly delineate counting area for accurate and precise results. Molded from a single piece of thermal and shock resistant glass with a polished chamber and cover glass assuring precise capillary loading and even cell distribution. Fluid volume is 0.1 mm3/mm2 and 0.00025 mm2 per each square.
Delivery information: The complete set includes one counting chamber, two 0.4 mm cover glass, one red cell pipette, one white cell pipette, tubing, mouthpieces, and storage case.
Specifications
  • Description:
    Hemacytometer Cover Glass, 0.4 mm (1461)