Orders LinkContactUsLinkComponent
Diamond Points, Electron Microscopy Sciences
Diamond Points, Small, 1.3 mm, Round
  100497-970
 :  62155-13
Diamond Points, Electron Microscopy Sciences
  100497-970
Diamond Points, Electron Microscopy Sciences
Diamond Points, Electron Microscopy Sciences
Diamond Points, Small, 1.3 mm, Round
$33.80
Each
All diamond points are mounted on 2.35 mm (3/32”) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
  • Ø:
    1.3 mm
  • Description:
    Diamond Points, Small
  • Shape:
    Round